Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter Chemistry Calibrator

Catalog No. 21280149 Shop All Beckman Coulter Products
Change view
Click to view available options
For Use With (Equipment):
DxC 500AU/AU480/AU680/DxC 700AU/AU5800/Beckman Coulter Analyzers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
21-280-149 DxC 500AU/AU480/AU680/DxC 700AU/AU5800/Beckman Coulter Analyzers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-280-149 Supplier Beckman Coulter Supplier No. 16550
Add to Cart
Add to Cart
This item is not returnable. View return policy

Our broad and growing portfolio of high-performing Six Sigma assays is compatible with AU Chemistry Analyzers and offers exceptional clinical quality.

  • Liquid ready-to-use reagents
  • Trust results with standardized test menus, assay protocols and reference ranges providing commutable patient results across AU analyzers
  • Select the right size for test volumes
  • Reagent Volume: D x C 500 AU/AU480/AU680/D x C 700 AU - 40 μL, AU5800 - 30 μL
TRUSTED_SUSTAINABILITY

Specifications

Detectable Analytes Albumin
Format Lyophilized
For Use With (Equipment) DxC 500AU/AU480/AU680/DxC 700AU/AU5800/Beckman Coulter Analyzers
Sample Type Serum, Plasma
Type Calibrator
Disposable Limited-use
Stability 28 Days
Sterility Sterile
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.