Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Exothermic/Endothermic Combination Demonstration
GREENER_CHOICESCI_ED

Catalog No. S96738 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S96738 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S96738 Supplier Innovating Science™ Supplier No. IS7006
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Study both processes of endothermic and exothermic reactions. Reactions are safely carried out in plastic bags. The students can pass the bags around to feel the results of the reactions.

Aligned to the Next Generation Science Standards (NGSS) Disciplinary Core Ideas: PS1.A; PS2.B Performance Expectations: MS-PS1-2 Cross Cutting Concepts: Patterns Engineering Practices: Analyzing and Interpreting Data Combo kit includes both IS7004 and IS7005 kits.

Specifications

Grade 11 to 14
For Use With (Application) Study both processes of endothermic and exothermic reactions
Includes 25 g iron metal powder, 1 g sodium chloride crystals, 5 g calcium chloride flakes, 1 tbsp vermiculite, 5 bags x 25 g ammonium nitrate un1942 + 1 tbsp vermiculite
Class Size 15 Lab Groups
Product Type Exothermic/Endothermic Combination Demonstration Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.