Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Foam City: The Catalytic Decomposition Of Hydrogen Peroxide Demonstration Kit
GREENER_CHOICESCI_ED

Catalog No. S96747 Shop All Aldon Products
Change view
Click to view available options
Class Size:
3 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S96747 3 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S96747 Supplier Innovating Science™ Supplier No. IS7019
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A large quantity of foam is produced when detergent and potassium iodide are added to hydrogen peroxide.

Aligned to the Next Generation Science Standards (NGSS) Disciplinary Core Ideas: PS1.A; PS1.B Performance Expectations: MS-PS1-2 Cross Cutting Concepts: Patterns Engineering Practices: Analyzing and Interpreting Data Kit Includes: 3 x 30 mL Hydrogen Peroxide, 30% 1 x 30 mL Dish Detergent, Green 1 x 10 g Potassium Iodide, Crystals

Specifications

Grade 11 to 14
Includes 3 x 30 mL hydrogen peroxide, 30% un2014, 1 x 30 mL dish detergent, green, 1 x 10 g potassium iodide, crystals
Class Size 3 Lab Groups
Product Type Blue Bottle Reaction Chemical Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.