Learn More
BURKLE INC Chemistry scoop, PP, volume 8.45 oz. (250 ml), rod 39.37 in. (100 cm) Ø 0.79 in. (20 mm)
Catalog No. 5000025696
Encompass_Preferred
Scoop for aggressive liquids, complete with strong handle, entirely made of polypropylene.
Catalog No. 50-000-25696 Supplier BURKLE INC Supplier No. 53530250
Scoop for aggressive liquids, complete with strong handle, entirely made of polypropylene.