Learn More
BURKLE INC Chemistry scoop, PP, volume 21.12 oz. (600 ml), rod 39.37 in. (100 cm) Ø 0.79 in. (20 mm)
Catalog No. 5000025749
Encompass_Preferred
Scoop for aggressive liquids, complete with strong handle, entirely made of polypropylene.
Catalog No. 50-000-25749 Supplier BURKLE INC Supplier No. 53530500
Scoop for aggressive liquids, complete with strong handle, entirely made of polypropylene.