Learn More
MSC Rochester Midland Corporation Proportioner, Regulator Knob Adjustment, Hose Inlet, Plastic Body
Catalog No. 502839961
Encompass
This proportioner is designed for accurate dispensing of liquids, featuring a regulator knob for precise adjustments. It supports both low and high flow applications and is compatible with EZ-Mix products and bottles.
  • Features 4 products
  • Includes a locking cabinet
  • Supports low flow applications
  • Supports high flow applications
  • Comes with 4 mating caps for EZ-Mix bottles
  • Enables both bucket and bottle filling
Catalog No. 50-283-9961 Supplier MSC Supplier No. 90709320
This proportioner is designed for accurate dispensing of liquids, featuring a regulator knob for precise adjustments. It supports both low and high flow applications and is compatible with EZ-Mix products and bottles.
  • Features 4 products
  • Includes a locking cabinet
  • Supports low flow applications
  • Supports high flow applications
  • Comes with 4 mating caps for EZ-Mix bottles
  • Enables both bucket and bottle filling