Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemizorb™ Granules

Catalog No. M1015681000 Shop All MilliporeSigma Products
Change view
Click to view available options
Packaging Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
M1015681000 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1015681000 Supplier MilliporeSigma™ Supplier No. 1015681000

Used as an absorbent for spilled liquids

  • Used as absorbent for liquid spills in laboratory
TRUSTED_SUSTAINABILITY

Specifications

Color White to colored
Type Absorbent
Format Granules
For Use With (Application) Laboratory cleaning
Packaging Quantity 1 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.