Learn More
Sterlitech Corporation Chemker 300 PTFE Vacuum Pump, 110V/60Hz
SDP
Catalog No. 501952306
Encompass_Preferred
Horse Power: 1/8, Flow Rate (lpm): 20High chemical resistance:All wetted parts of Chemker series pump are made of PTFE material which can resist most of corrosive gases.Applications :* Vacuum filtration* Gel electrophoresis * Solid-Phase Extraction (SPE)* Solvent purification
Catalog No. 50-195-2306 Supplier Sterlitech Corporation Supplier No. 16930011
Horse Power: 1/8, Flow Rate (lpm): 20High chemical resistance:All wetted parts of Chemker series pump are made of PTFE material which can resist most of corrosive gases.Applications :* Vacuum filtration* Gel electrophoresis * Solid-Phase Extraction (SPE)* Solvent purification