Learn More
Sterlitech Corporation Chemker 400 PTFE Vacuum Pump, 110V/60Hz
SDP
Catalog No. 501952307
Encompass_Preferred
Horse Power: 1/6, Flow Rate (lpm): 38High chemical resistance:All wetted parts of Chemker series pump are made of PTFE material which can resist most of corrosive gases.Applications :* Vacuum filtration* Gel electrophoresis * Solid-Phase Extraction (SPE)* Solvent purification
Catalog No. 50-195-2307 Supplier Sterlitech Corporation Supplier No. 16940011
Horse Power: 1/6, Flow Rate (lpm): 38High chemical resistance:All wetted parts of Chemker series pump are made of PTFE material which can resist most of corrosive gases.Applications :* Vacuum filtration* Gel electrophoresis * Solid-Phase Extraction (SPE)* Solvent purification