Learn More
Electron Microscopy Sciences Chemkit for Preparations of Biological Specimens
SDP
Catalog No. 50980439
Encompass_Preferred
This package is arranged to enable a laboratory to perform standard preparations of biological specimens with enough supplies to last. Includes:
Fixatives-osmium tetroxide crystal, osmium tetroxide 4% solution, glutaraldehyde EM grade 50%, glutaraldehyde EM grade 8%, and paraformaldehyde Prill.
Buffers-sodium phosphate monobasic, sodium phosphate dibasic, sodium hydroxide pellets, and sodium cacodylate trihydrate.
Dehydrants-propylene oxide and acetone.
Embedding media-EMBed-812 kit and low viscosity kit (Dr. Spurr).
Stains-toluidine blue, borax tetra, sodium borate, uranyl acetate, lead citrate, and epoxy tissue stain.
Catalog No. 50-980-439 Supplier Electron Microscopy Sciences Supplier No. 14750
This package is arranged to enable a laboratory to perform standard preparations of biological specimens with enough supplies to last. Includes:
Fixatives-osmium tetroxide crystal, osmium tetroxide 4% solution, glutaraldehyde EM grade 50%, glutaraldehyde EM grade 8%, and paraformaldehyde Prill.
Buffers-sodium phosphate monobasic, sodium phosphate dibasic, sodium hydroxide pellets, and sodium cacodylate trihydrate.
Dehydrants-propylene oxide and acetone.
Embedding media-EMBed-812 kit and low viscosity kit (Dr. Spurr).
Stains-toluidine blue, borax tetra, sodium borate, uranyl acetate, lead citrate, and epoxy tissue stain.