Learn More
Med Vet International Chemo Dispensing Pins™ w/ Security Clip, J0463V, Each
Catalog No. 5000581694
Encompass_Preferred
Chemo Dispensing Pins™ designed to eliminate aerosoling of chemotherapeutic medications during the reonstitution and dispensing process. Assists in USP compliance. DEHP-free. Latex-free. Chemo Dispensing Pin™ with security clip for attachment to standard 20mm rubber-stoppered vials. Built in 0.2 micron hydrophobic air-venting filter. Proximal luer lock connector.
Catalog No. 50-005-81694 Supplier Med Vet International Supplier No. JOR463V
Chemo Dispensing Pins™ designed to eliminate aerosoling of chemotherapeutic medications during the reonstitution and dispensing process. Assists in USP compliance. DEHP-free. Latex-free. Chemo Dispensing Pin™ with security clip for attachment to standard 20mm rubber-stoppered vials. Built in 0.2 micron hydrophobic air-venting filter. Proximal luer lock connector.