Learn More
Cusabio Technology LLC chemokine C-C motif ligand 17
Supplier:  Cusabio Technology LLC CSBE09257H
Encompass_Preferred
This kit detects CCL17 (Human thymus activation regulated chemokine/TARC) in serum plasma and tissue homogenates Designed for quantitative analysis it employs a sandwich ELISA method with a detection range of 15 6 pg/mL to 1000 pg/mL and a sensitivity of 3 9 pg/mL ensuring accurate measurement of low-concentration samples The assay can be completed within 1 to 5 hours using 50-100 L of sample per test with absorbance readings taken at 450 nm Suitable for both research and diagnostic applications this kit provides a reliable and efficient solution for quantifying TARC/CCL17 levels in diverse biological samples
Catalog No. 50-000-50382