Learn More
Cusabio Technology LLC chemokine C-C motif ligand 3
Supplier:  Cusabio Technology LLC CSBE07422R
Encompass_Preferred
This kit detects CCL3 (macrophage inflammatory protein 1A MIP-1A) in rat serum plasma and tissue homogenates using a quantitative sandwich ELISA method It offers a detection range of 12 5 pg/mL to 800 pg/mL with a sensitivity of 3 12 pg/mL ensuring precise measurement of low-abundance targets The assay requires a sample volume of 50-100 L and delivers results within 1-5 hours Absorbance is measured at 450 nm providing reliable data for research applications in inflammation and immune response studies Designed for accuracy and efficiency this kit is optimized for reproducible performance across biological matrices supporting both basic research and preclinical investigations
Catalog No. 50-000-50393