Learn More
Cusabio Technology LLC chemokine C-C motif ligand 4
Catalog No. 5000051534
Encompass_Preferred
This kit detects CCL4 (Macrophage Inflammatory Protein 1B MIP-1B) in mouse serum plasma and tissue homogenates using a quantitative sandwich ELISA method It offers a detection range of 12 5 pg/mL to 800 pg/mL with a sensitivity of 3 12 pg/mL ensuring accurate measurement across biological samples The assay requires 50-100 L of sample per test and involves incubation steps spanning 1 to 5 hours Absorbance is measured at 450 nm for quantification providing a robust tool for studying inflammatory biomarkers in preclinical research Designed for specificity and reproducibility this kit is ideal for applications in immunology and cellular response studies involving mouse models
Catalog No. 50-000-51534 Supplier Cusabio Technology LLC Supplier No. CSBE04665M
This kit detects CCL4 (Macrophage Inflammatory Protein 1B MIP-1B) in mouse serum plasma and tissue homogenates using a quantitative sandwich ELISA method It offers a detection range of 12 5 pg/mL to 800 pg/mL with a sensitivity of 3 12 pg/mL ensuring accurate measurement across biological samples The assay requires 50-100 L of sample per test and involves incubation steps spanning 1 to 5 hours Absorbance is measured at 450 nm for quantification providing a robust tool for studying inflammatory biomarkers in preclinical research Designed for specificity and reproducibility this kit is ideal for applications in immunology and cellular response studies involving mouse models