Learn More
Cusabio Technology LLC chemokine C-C motif ligand 8
Catalog No. 5000050539
Encompass_Preferred
This ELISA kit is designed for the quantitative detection of Mouse monocyte chemotactic protein 2 (MCP-2/CCL8) in serum plasma and tissue homogenates using a sandwich assay format With a detection range of 6 25 pg/mL to 400 pg/mL and a sensitivity of 1 56 pg/mL it provides precise measurements for low-abundance samples The assay requires 50-100 L of sample per test and delivers results within 1-5 hours through absorbance measurement at 450 nm Its standardized protocol ensures reliable performance in research applications requiring accurate quantification of CCL8 chemokine levels across biological matrices
Catalog No. 50-000-50539 Supplier Cusabio Technology LLC Supplier No. CSBE07428M
This ELISA kit is designed for the quantitative detection of Mouse monocyte chemotactic protein 2 (MCP-2/CCL8) in serum plasma and tissue homogenates using a sandwich assay format With a detection range of 6 25 pg/mL to 400 pg/mL and a sensitivity of 1 56 pg/mL it provides precise measurements for low-abundance samples The assay requires 50-100 L of sample per test and delivers results within 1-5 hours through absorbance measurement at 450 nm Its standardized protocol ensures reliable performance in research applications requiring accurate quantification of CCL8 chemokine levels across biological matrices