Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chemokine (C-X-C motif) ligand 13 (CXCL13) | >95.0% | 2 UG
SDP

Catalog No. 5000293905
Encompass_Preferred

Recombinant cynomolgus CXCL13 (BCA-1) is a chemokine that selectively chemoattracts B lymphocytes via the CXCR5 receptor. Supplied as a lyophilized recombinant protein expressed in E. coli with an N-terminal His tag, it is intended for research use in chemotaxis, receptor binding, and functional assays.

  • Produced in E. coli with an N-terminal His tag.
  • Lyophilized from 10 mM Tris, 250 mM NaCl, pH 8.0.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Endotoxin <1 EU/μg (LAL method).
  • Biological activity: ED50 0.019 μg/mL in Ramos cell chemotaxis assay.
  • Appearance: lyophilized powder; reconstitute and aliquot for storage.

Catalog No. 50-002-93905 Supplier Medchemexpress LLC Supplier No. HYP761642UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant cynomolgus CXCL13 (BCA-1) is a chemokine that selectively chemoattracts B lymphocytes via the CXCR5 receptor. Supplied as a lyophilized recombinant protein expressed in E. coli with an N-terminal His tag, it is intended for research use in chemotaxis, receptor binding, and functional assays.

  • Produced in E. coli with an N-terminal His tag.
  • Lyophilized from 10 mM Tris, 250 mM NaCl, pH 8.0.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Endotoxin <1 EU/μg (LAL method).
  • Biological activity: ED50 0.019 μg/mL in Ramos cell chemotaxis assay.
  • Appearance: lyophilized powder; reconstitute and aliquot for storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.