Learn More
Meso Scale Discovery U-PLEX Chemokine Combo 2 Mouse, Quantitative multiplex immunoassay kit, Detects 6 analytes, Small volume requirement, High sensitivity

Supplier: Meso Scale Discovery K15319K2
The MESO SCALE DISCOVERY U-PLEX Chemokine Combo 2 (mouse) is a sensitive multiplex immunoassay kit for measuring key mouse chemokines. It utilizes MSD's MULTI-ARRAY technology, enabling rapid detection of multiple targets using a small sample volume. The assay delivers detection limits in the low pg/mL range and shows minimal background signals for accurate results across various research applications.
- Product type: Multiplex immunoassay kit
- Species reactivity: Mouse
- Analytes detected: 6CKine/CCL21, BCA-1/BLC, MCP-5/CCL12, RANTES, SDF-1α, TARC
- Detection technology: MSD MULTI-ARRAY, U-PLEX platform
- Sensitivity: Sub- to low pg/mL range
- Multiplexing capability: True
- Sample volume requirement: Small volume
- Dynamic range: Wide
- Background signal: Minimal
- Plate read time: 90 seconds
- Application: Quantitative biomarker measurement
- Regulatory status: For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.