Learn More
Cayman Chemical ChemokineLike Receptor 1 Bloc
SDP
Supplier:  Cayman Chemical 103261 EA
Encompass_Preferred
To be used in conjunction with Cayman’s Chemokine-Like Receptor 1 Polyclonal Antibody to block protein-antibody complex formation during immunochemical analysis of Chemokine-Like Receptor 1
Catalog No. 50-288-6578