Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cardinal Health™ ChemoPlus™ Non-Sterile Poly-Coated Full Coverage Chemotherapy Gowns, Full Back

Catalog No. 17009983 Shop All Cardinal Health Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-009-983 2X-Large
17-009-980 Medium
17-009-981 Large
17-009-982 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 17-009-983 Supplier Cardinal Health™ Supplier No. CT5505T

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Non-sterile gowns protect healthcare personnel from exposure to toxic chemotherapy drugs during preparation, handling, and administration.

  • AAMI Level 3
  • Evaluated for resistance to permeation of 16 chemotherapy drugs per ASTM F739-20
  • Meet USP <800> standards for handling hazardous drugs
  • Reinforced critical zones include chest, sleeve seam, sleeve and front belt attachment points where direct contact with infectious material is most likely to occur
  • Full back style gowns that close and tie in the back
  • Hoop and loop neck, waist ties

Specifications

Length (English) 51.1 in.
Length (Metric) 129.794 cm
Width (English) 33.5 in.
Width (Metric) 85.09 cm
Gender Unisex
No. of Pockets None
For Use With (Application) Hazards associated with handling chemotherapy drugs
Product Type Chemotherapy Gown
Size 2X-Large
Material Polyethylene-coated Polypropylene
Color Blue
Closure Type Waist Ties
Disposable Single-use
Sterility Non-sterile
Cuff Style Knit
Model CT5505T
Sleeve Style Long Sleeve
Certifications/Compliance AAMI Level 3, ASTM F739-20, USP 800 Standards, FDA 510K
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.