Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cardinal Health™ ChemoPlus™ Non-Sterile Poly-Coated Full Coverage Chemotherapy Gowns, Open Back

Catalog No. 17009985 Shop All Cardinal Health Products
Change view
Click to view available options
Size:
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-009-985 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-009-985 Supplier Cardinal Health™ Supplier No. DP5001GT
This item is not returnable. View return policy

Non-sterile gowns protect healthcare personnel from exposure to toxic chemotherapy drugs during preparation, handling, and administration.

  • AAMI Level 3
  • Evaluated for resistance to permeation of 16 chemotherapy drugs per ASTM F739-20
  • Meet USP <800> standards for handling hazardous drugs
  • Reinforced critical zones include chest, sleeve seam, sleeve and front belt attachment points where direct contact with infectious material is most likely to occur
  • Open back gowns that provide front coverage and breathability
  • Hoop and loop neck, waist ties

Specifications

Width (English) 24.75 in.
Length (English) 46.5 in.
Width (Metric) 62.86 cm
Length (Metric) 118.11 cm
Size Large
Gender Unisex
No. of Pockets None
Product Type Chemotherapy Gown
For Use With (Application) Hazards associated with handling chemotherapy drugs
Material Polyethylene-coated Polypropylene
Color Blue
Closure Type Waist Ties
Disposable Single-use
Sterility Non-sterile
Cuff Style Knit
Model DP5001GT
Sleeve Style Long Sleeve
Certifications/Compliance AAMI Level 3, ASTM F739-20, USP 800 Standards, FDA 510K
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.