Learn More
BURKLE INC ChemoSampler, PP grey, 33.81 oz. (1000 ml), 39.37 in. (100 cm)
Catalog No. 5000025693
Encompass_Preferred
The samples are drawn directly into the screw-on original sampling bottle. The tamperproof bottle closure of these bottles with original safety ring ensures unadulterated sample transport. Samples can be taken from depths of as much as 200 cm (78.74 in.), e.g. from tanks, silos or tankers, by using the extension rod.
Catalog No. 50-000-25693 Supplier BURKLE INC Supplier No. 53361000
The samples are drawn directly into the screw-on original sampling bottle. The tamperproof bottle closure of these bottles with original safety ring ensures unadulterated sample transport. Samples can be taken from depths of as much as 200 cm (78.74 in.), e.g. from tanks, silos or tankers, by using the extension rod.