Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Chemotactic Peptide

Catalog No. AAJ65184MB
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65184MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ65184MB Supplier Thermo Scientific Chemicals Supplier No. J65184MB
Only null left
Add to Cart

437.55 g/mol

Induces leucocyte chemotaxis, and activates macrophages

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Purification Method HPLC
Common Name fMLP
Content And Storage -30°C to -10°C
Format Lyophilized Powder
For Use With (Application) Induces leucocyte chemotaxis, and activates macrophages
Molecular Weight (g/mol) 437.55
Product Type Chemotactic Peptide
Quantity 25 mg
Regulatory Status RUO - research use only
Sequence N-Formyl-Met-Leu-Phe

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.