Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Chemotaxis: The Science of Attraction in C. elegans Experiment Kit
SDPSCI_ED

Catalog No. S29025 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S29025 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S29025 Supplier Edvotek™ Supplier No. 852
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

In this experiment, students observe and record the phenomenon by which normal and mutant strains of C. elegans can direct their movement in response to certain chemicals in the environment. Edvotek™ Chemotaxis: The Science of Attraction in C. elegans Experiment Kit contains five LIVE materials.

  • For 10 Lab Groups
  • Growing Bacteria: overnight; Plating Worms: 15 min.; Worm Growth: 3-4 Days; Toxicity Experiment: 45-60 min.

Specifications

Grade 9 to Undergraduate
Includes Instructions, C. elegans - normal, C. elegans Chemotaxis - mutant, petri dishes, NGM medium, E. coli OP50 BactoBeads™, cell counting chambers, buffer, pipettes, sterile loops, tubes and chemical compounds.
Time Range Growing Bacteria: overnight; Plating Worms: 15 min.; Worm Growth: 3-4 Days; Chemotaxis: 45-60 min.
Content And Storage Some Components Require Refrigerator Storage
Class Size 10 Lab Groups
Product Type Chemotaxis: The Science of Attraction in C. elegans Experiment Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.