Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Chemotherapy Sharps Collectors

Catalog No. BD305604 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Capacity (English):
5 gal.
9 gal.
17 gal.
Closure Type:
Hinged Top
Plug Cap
Slide Top
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Closure Type
BD305604 9 gal. Slide Top
14-826-118 17 gal. Hinged Top
14-826-131 9 gal. Hinged Top
14-826-95B 5 gal. Plug Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. BD305604 Supplier BD Supplier No. 305604
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Wide entry ports design for disposal of bulky chemotherapy waste

  • Designed to offer increased leak resistance
  • Cannot be autoclaved
  • Yellow
  • Collectors are marked with biohazard warning label

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 34 L
Depth (English) 11.7 in.
Closure Type Slide Top
Depth (Metric) 29.7 cm
Closure Color Clear
Capacity (English) 9 gal.
Color Yellow
Height (Metric) 47.6 cm
Latex Free Yes
Sterility Non-sterile
Width (Metric) 44.9 cm
Autoclavable Not Autoclavable
Includes Inner gasket lining, biohazard warning label
For Use With (Application) For disposal of trace chemotherapy waste
Width (English) 17.7 in.
Height (English) 18.75 in.
Material Polypropylene
Product Type Chemotherapy Sharps Collector
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.