Learn More
Ace Glass, Inc. CHEMRAZ O-RING FOR NO36 CHEMRAZ O-RING FOR NO36
SDP
Catalog No. NC0021451
Encompass
Manufactured by Greene, Tweed & Company and Supplied by Ace Glass Inc. Chemraz 514 A is a white perfluoroelastomer with the broadest chemical resistance of any elestomeric material. Avoid acids, aldehydes, ethylene oxide and propylene oxide. Temp range -30 to 220 deg.C
Catalog No. NC0021451 Supplier Ace Glass, Inc. Supplier No. 7859572
Manufactured by Greene, Tweed & Company and Supplied by Ace Glass Inc. Chemraz 514 A is a white perfluoroelastomer with the broadest chemical resistance of any elestomeric material. Avoid acids, aldehydes, ethylene oxide and propylene oxide. Temp range -30 to 220 deg.C