Learn More
Ace Glass, Inc. CHEMRAZ O-RING, SIZE 111
SDP
Catalog No. NC0626473
Encompass
Manufactured by Greene, Tweed & Company and Supplied by Ace Glass Inc. Molded of perfluoroelastimer polymer, CHEMRAZ has the broadest chem- ical resistance of any elastomeric material. Combines the resiliance and sealing force of an elastomer with chemical resistance approaching that of PTFE. Resistant to swelling and embrittlement, easily in- stalled, conforming, not likely to creep or cold flow.
Catalog No. NC0626473 Supplier Ace Glass, Inc. Supplier No. 7859518
Manufactured by Greene, Tweed & Company and Supplied by Ace Glass Inc. Molded of perfluoroelastimer polymer, CHEMRAZ has the broadest chem- ical resistance of any elastomeric material. Combines the resiliance and sealing force of an elastomer with chemical resistance approaching that of PTFE. Resistant to swelling and embrittlement, easily in- stalled, conforming, not likely to creep or cold flow.