Learn More
Ace Glass, Inc. CHEMRAZ WHITE O-RINGS 009 CHEMRAZ WHITE O-RINGS 009
SDP
Catalog No. NC9194340
Encompass
Manufactured by Greene, Tweed & Company and Supplied by Ace Glass Inc. O-Ring, Chemraz, -009, Molded of Perfluoroelastimer Polymer, Chemraz has Broadest Chemical Resistance of any Elastomeric Material, Resistant to Swelling and Embrittlement, Easily in- Stalled, Conforming, Not Likely to Creep or Cold Flow
Catalog No. NC9194340 Supplier Ace Glass, Inc. Supplier No. 7859507
Manufactured by Greene, Tweed & Company and Supplied by Ace Glass Inc. O-Ring, Chemraz, -009, Molded of Perfluoroelastimer Polymer, Chemraz has Broadest Chemical Resistance of any Elastomeric Material, Resistant to Swelling and Embrittlement, Easily in- Stalled, Conforming, Not Likely to Creep or Cold Flow