Learn More
Chemglass Life Sciences 300mL ChemRxnHub Process System Complete with Glassware, Stand, and OHS 100 Advance Motor
SDP
Catalog No. 50143623
Encompass_Preferred
ChemRxnHub™ process systems are complete with benchtop stand, overhead stirrer and glassware. Available with the optional manifold connection and heat transfer fluid draining system. The unique design of the ChemRxnHub™ support stand and clamping system allows for quick and easy installation/removal of glassware and fits into most benchtop hoods.
Catalog No. 50-143-623 Supplier Chemglass Life Sciences Supplier No. CG1949X300
Add to Cart
Add to Cart
Request a Quote
ChemRxnHub™ process systems are complete with benchtop stand, overhead stirrer and glassware. Available with the optional manifold connection and heat transfer fluid draining system. The unique design of the ChemRxnHub™ support stand and clamping system allows for quick and easy installation/removal of glassware and fits into most benchtop hoods.