Learn More
Chemglass Life Sciences ChemRxnHub Support Stand with Manifold System
SDP
Catalog No. 501531294
Encompass_Preferred
ChemRxnHub Support Stand with Manifold System. Includes (1) Reactor Support Frame, (1) S.S Reactor Clamp, 100mm, (1) S.S. Reactor Clamp, 150mm, (1) Outlet Manifold Assembly Complete, (1) Inlet Manifold Assembly Complete. This stationary reactor stand is designed to fit into most benchtop hoods. Stand can hold a single reactor. The stand provides support for the Reaction Vessel(s), Glassware, Overhead Stirring Motor, & Temperature Monitor. Stand includes removable spill tray. Weight of stand is 44lbs. S.S. Uprights&Heavy Duty PTFE Coated Aluminum Fittings Provide Max Strength & Chemical Durability. Supports Reactors from 300mL to 5L. Removable Polyethylene Spill Tray. Middle Support Bracket is Vertically Adjustable. It Accepts 100 & 150mm S.S. Support Clamp. Adjustable Top Support Bracket is Vertically Adjustable. Mounting Clamps Accept Overhead Stirrers with a MAX Support Arm of 5/8 Diameter. Frame has Adjustable Leveling Pads. Compact Foot-Print: 15.25 W x 17.25 D x 44 H.
Catalog No. 50-153-1294 Supplier Chemglass Life Sciences Supplier No. CG1949X105M
Add to Cart
Add to Cart
Request a Quote
ChemRxnHub Support Stand with Manifold System. Includes (1) Reactor Support Frame, (1) S.S Reactor Clamp, 100mm, (1) S.S. Reactor Clamp, 150mm, (1) Outlet Manifold Assembly Complete, (1) Inlet Manifold Assembly Complete. This stationary reactor stand is designed to fit into most benchtop hoods. Stand can hold a single reactor. The stand provides support for the Reaction Vessel(s), Glassware, Overhead Stirring Motor, & Temperature Monitor. Stand includes removable spill tray. Weight of stand is 44lbs. S.S. Uprights&Heavy Duty PTFE Coated Aluminum Fittings Provide Max Strength & Chemical Durability. Supports Reactors from 300mL to 5L. Removable Polyethylene Spill Tray. Middle Support Bracket is Vertically Adjustable. It Accepts 100 & 150mm S.S. Support Clamp. Adjustable Top Support Bracket is Vertically Adjustable. Mounting Clamps Accept Overhead Stirrers with a MAX Support Arm of 5/8 Diameter. Frame has Adjustable Leveling Pads. Compact Foot-Print: 15.25 W x 17.25 D x 44 H.