Learn More
International Enviroguard™ ChemSplash 1 Chemical Resistant Apron with Ties in Back
Catalog No. 17184104 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Closure Type:
Waist Ties
Catalog No. Closure Type
17-184-104 Waist Ties
1 options
Catalog No. 17-184-104 Supplier International Enviroguard™ Supplier No. 702846Y
Add to Cart
Add to Cart
This chemical-resistant apron provides frontal protection against chemical splashes and toxic particles.
  • Longer length ensures your chest and knees are protected from light splashes and sprays
  • Proprietary, hybrid membrane fabric helps keep you cool as you work long hours in order to prevent heat stress
  • Tie strings are secured in the back so they're out of the way while you work, also allowing for a more adjustable fit
  • Comfortable bib-style design
  • Possible applications include manufacturing, emergency response, chemical mixing, pesticide application, environmental cleanup, and more
Specifications
Type Protective Apron
Size Universal
Material Multi-layer Barrier Laminate Fabric
Color Yellow
Disposable Single-use
Closure Type Waist Ties
Sterility Non-sterile
Latex Free Yes
No. of Pockets None
Length (English) 46 in.
Width (English) 28 in.
Seam Type Seamless