Learn More
International Enviroguard™ ChemSplash 1 Chemical Splash Coveralls with Taped Seams, Elastic Wrist and Ankle
Catalog No. 17184132 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
X-Large
Catalog No. Size
17-184-132 X-Large
1 options
Catalog No. 17-184-132 Supplier International Enviroguard™ Supplier No. 7013YTXL
Only null left
Add to Cart
Add to Cart
These durable and disposable coveralls with taped seams and elastic back protect against chemical splashes and toxic particles.
  • Proprietary, hybrid membrane fabric offers a high moisture vapor transfer rate (MVTR), so you stay cool as you put in a hard day's work
  • A storm flap over the zipper with pressure-sensitive tape helps prevent hazardous liquids from seeping through exposed zipper seams
  • A high-contrast red stripe lets you quickly distinguish this taped-seam garment from coveralls made with serged seams
  • For added comfort, seamless shoulders help prevent tears and allow you to move more freely as you work
  • Roomy, seamless shoulder area resists tears and provides more protection in key exposure area
  • A wider, gusseted crotch gives the wearer more room to move in all directions without ripping fabric
  • Elastic back stretches with user for more comfort and better tear resistance
  • Storm flap over zipper with pressure-sensitive tape adds an extra layer of protection
  • High moisture vapor transfer rate improves wearer comfort
  • Elastic wrists offer a secure fit to keep sleeves in place to prevent skin exposure
  • Possible applications include manufacturing, emergency response, environmental cleanup, and more
Specifications
Product Type Chemical Protective Clothing
Size X-Large
Gender Unisex
Material Laminated Non-woven Fabric
Color Yellow
Disposable Single-use
Waist Style Elastic at Back
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Standard Collar
Closure Type 2-way Front Zipper with Storm Flap
Seam Type Serged, Taped
Sterility Non-sterile
Resistance Acid, Caustics, Chemicals, Isopropyl Alcohol, Particle
Latex Free Yes
No. of Pockets None