Learn More
International Enviroguard™ ChemSplash 2 Chemical Splash Coveralls with Taped Seams, Attached Hood and Boots
Catalog No. 17184086 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
3X-Large
Catalog No. Size
17-184-086 3X-Large
1 options
Catalog No. 17-184-086 Supplier International Enviroguard™ Supplier No. 7319GT3XL
Add to Cart
Add to Cart
These durable and disposable coveralls with elastic wrists and back are designed to protect against a broad range of acids and caustic chemicals.
  • Proprietary fabric is lighter and more pliable than competitive coveralls, so you can work comfortably hour after hour
  • Taped seams (blue tape) provide ideal protection to coveralls made with serged seams that are exposed
  • With an attached hood and boots, you're protected from head to toe
  • Attached thumb loops keep sleeves in place while an enlarged zipper pull makes it easier to grab with gloved hands
  • An attached hood, boots, and elastic wrists provide full-body protection from chemical hazards
  • Light, pliable fabric for more comfortable wear
  • Elastic back stretches with user for more comfort and better tear resistance
  • Storm flap over the zipper with pressure-sensitive tape adds an extra layer of protection by sealing off points in a seam where liquids can penetrate (needle holes)
  • Possible applications include emergency response, chemical loading, petrochemical environments, and more
Specifications
Product Type Chemical Protective Clothing
Size 3X-Large
Gender Unisex
Material Multi-layer Barrier Laminate Fabric
Color Gray
Disposable Single-use
Waist Style Elastic at Back
Ankle Style Attached Boots
Cuff Style Elastic, Thumb Loop
Neck Style Hooded
Hood Type Attached, Elastic Face
Closure Type 2-way Zipper with Sealed Storm Flap
Seam Type Serged, Taped
Sterility Non-sterile
Resistance Acid, Bloodborne Pathogens, Chemicals, Isopropyl Alcohol, Caustics
Latex Free Yes
No. of Pockets None