Learn More
Chemtronics™ Coventry™ Poly-Wipes
Catalog No. 19061613
Change view
Click to view available options
Dimensions (L x W):
9 x 9 in.
Catalog No. Dimensions (L x W)
19-061613 9 x 9 in.
1 options
This item is not returnable. View return policy
Catalog No. 19-061613 Supplier Chemtronics™ Supplier No. 6209
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specifications
Material Polyester
Absorbancy Highly absorbent
Construction Monofilament
Edges Heat cut
Dimensions (L x W) 9 x 9 in.
For Use With (Application) Semiconductor Wafer Fabs; Aerospace Production Areas; Disk Drives Production Areas; Pharmaceutical / Biotechnical Production Areas; General Cleanroom Cleaning; Clean and polish critical surfaces both metal and non-metal
Packaging Quantity 150 Wipes/Bag