Learn More
Chemtronics™ Coventry™ Polyester Swab
Catalog No. 19083300
Change view
Click to view available options
Tip Style:
Flat
Catalog No. Tip Style
19-083-300 Flat
1 options
This item is not returnable. View return policy
Catalog No. 19-083-300 Supplier Chemtronics™ Supplier No. 38140
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specifications
Quantity 500
For Use With (Application) Cleans SFF connectors, alignment pins and sleeves; Cleans test equipment ports
Handle Color Gray
Packaging Bag
Tip Style Flat