Learn More
Chemtronics™ Flextips™ Swab
Catalog No. 19041347
Change view
Click to view available options
Tip Material:
Polyurethane Foam
Catalog No. Tip Material
19-041-347 Polyurethane Foam
1 options
Catalog No. 19-041-347 Supplier Chemtronics™ Supplier No. CX50
Specifications
Quantity 50
For Use With (Application) Clean excess adhesives after gluing Clean micro-mechanical devices; Remove Flux residues from printed circuit boards; Apply small precise quantities of adhesive or lubricants
Packaging Bag
Handle Material Polypropylene
Tip Material Polyurethane Foam