Learn More
Chemglass Life Sciences 0-8mm Chem-Vac Chem-Cap Valve
SDP
Catalog No. 501532007
Encompass_Preferred
0-8mm Chem-Vac Chem-Cap Valve. CHEM-VAC CHEM-CAP valve has a single sidearm sealed at a 90 angle to the barrel. This inexpensive configuration allows for the lower portion of the barrel to be sealed directly to the apparatus being constructed, making for more compact, stronger assemblies. See CG-961 for replacement CHEM-CAP plug and CG-902 for glass portion only.
Catalog No. 50-153-2007 Supplier Chemglass Life Sciences Supplier No. CGB96002
0-8mm Chem-Vac Chem-Cap Valve. CHEM-VAC CHEM-CAP valve has a single sidearm sealed at a 90 angle to the barrel. This inexpensive configuration allows for the lower portion of the barrel to be sealed directly to the apparatus being constructed, making for more compact, stronger assemblies. See CG-961 for replacement CHEM-CAP plug and CG-902 for glass portion only.