Learn More
Cayman Chemical ChenodeoxycholIc ACDodIM 500mg
SDP
Catalog No. 502914046
Encompass_Preferred
A primary bile acid; an FXR agonist that binds to FXRs in a TR-FRET assay (EC50 = 13 µM) and induces FXR transactivation in a reporter assay; induces transcription of GCL in primary hepatocytes and HepG2 cells from 25-100 µM
Catalog No. 50-291-4046 Supplier Cayman Chemical Supplier No. 35346500 MG
Add to Cart
Add to Cart
A primary bile acid; an FXR agonist that binds to FXRs in a TR-FRET assay (EC50 = 13 µM) and induces FXR transactivation in a reporter assay; induces transcription of GCL in primary hepatocytes and HepG2 cells from 25-100 µM