Learn More
Apexbio Technology LLC Chenodeoxycholic Acid 474-25-9 100mg
SDP
Catalog No. 5000089937
Encompass_Preferred
Chenodeoxycholic Acid is a small-molecule agonist targeting the farnesoid X receptor (FXR) It is designed to selectively activate FXR thereby regulating critical signaling pathways involved in bile acid homeostasis lipid metabolism and cholesterol regulation Chenodeoxycholic Acid exerts its biological activity primarily through FXR-dependent modulation of transcriptional pathways Based on these pharmacological properties Chenodeoxycholic Acid holds research potential in the investigation of hepatic metabolism mechanisms cholesterol balance metabolic disorders and FXR-mediated gene expression in various physiological and pathological conditions
Catalog No. 50-000-89937 Supplier Apexbio Technology LLC Supplier No. B1908100
Add to Cart
Add to Cart
Chenodeoxycholic Acid is a small-molecule agonist targeting the farnesoid X receptor (FXR) It is designed to selectively activate FXR thereby regulating critical signaling pathways involved in bile acid homeostasis lipid metabolism and cholesterol regulation Chenodeoxycholic Acid exerts its biological activity primarily through FXR-dependent modulation of transcriptional pathways Based on these pharmacological properties Chenodeoxycholic Acid holds research potential in the investigation of hepatic metabolism mechanisms cholesterol balance metabolic disorders and FXR-mediated gene expression in various physiological and pathological conditions