Learn More
Apexbio Technology LLC Chenodeoxycholic Acid 474-25-9 1g
SDP
Catalog No. 5000089936
Encompass_Preferred
Chenodeoxycholic Acid is a small-molecule agonist targeting the farnesoid X receptor (FXR) It is designed to selectively activate FXR thereby regulating critical signaling pathways involved in bile acid homeostasis lipid metabolism and cholesterol regulation Chenodeoxycholic Acid exerts its biological activity primarily through FXR-dependent modulation of transcriptional pathways Based on these pharmacological properties Chenodeoxycholic Acid holds research potential in the investigation of hepatic metabolism mechanisms cholesterol balance metabolic disorders and FXR-mediated gene expression in various physiological and pathological conditions
Catalog No. 50-000-89936 Supplier Apexbio Technology LLC Supplier No. B19081000
Chenodeoxycholic Acid is a small-molecule agonist targeting the farnesoid X receptor (FXR) It is designed to selectively activate FXR thereby regulating critical signaling pathways involved in bile acid homeostasis lipid metabolism and cholesterol regulation Chenodeoxycholic Acid exerts its biological activity primarily through FXR-dependent modulation of transcriptional pathways Based on these pharmacological properties Chenodeoxycholic Acid holds research potential in the investigation of hepatic metabolism mechanisms cholesterol balance metabolic disorders and FXR-mediated gene expression in various physiological and pathological conditions