Learn More
Medchemexpress LLC Chenodeoxycholic Acid | 474-25-9 | 99.9% | C24H40O4 | 1 ML
SDP
Catalog No. 5000433969
Encompass_Preferred
Chenodeoxycholic Acid is a hydrophobic primary bile acid that activates nuclear receptors (FXR) involved in cholesterol metabolism. It is for research use only.
  • Endogenous metabolite.
  • Activates nuclear receptors (FXR) involved in cholesterol metabolism.
Catalog No. 50-004-33969 Supplier Medchemexpress LLC Supplier No. HY7684710MM/1ML
Add to Cart
Add to Cart
Chenodeoxycholic Acid is a hydrophobic primary bile acid that activates nuclear receptors (FXR) involved in cholesterol metabolism. It is for research use only.
  • Endogenous metabolite.
  • Activates nuclear receptors (FXR) involved in cholesterol metabolism.