Learn More
Medchemexpress LLC Chenodeoxycholic acid | 474-25-9 | MFCD00064142 | 99.8% | 392.57 g/mol | C24H40O4 | 10 MG
SDP
Catalog No. 5000298725
Encompass_Preferred
Chenodeoxycholic acid is an analytical reference standard of a hydrophobic primary bile acid (CDCA) used in research and analytical assays related to bile acid and cholesterol metabolism. Supplied at high purity for reliable calibration and quantitative analysis.
  • High purity (99.80%) suitable for analytical applications.
  • Provided in small pack sizes for standards and method development.
  • Used as a reference compound in bile acid and cholesterol metabolism studies.
  • Applicable for calibration, quantitative assays, and method validation.
Catalog No. 50-002-98725 Supplier Medchemexpress LLC Supplier No. HY76847R10MG
Add to Cart
Add to Cart
Chenodeoxycholic acid is an analytical reference standard of a hydrophobic primary bile acid (CDCA) used in research and analytical assays related to bile acid and cholesterol metabolism. Supplied at high purity for reliable calibration and quantitative analysis.
  • High purity (99.80%) suitable for analytical applications.
  • Provided in small pack sizes for standards and method development.
  • Used as a reference compound in bile acid and cholesterol metabolism studies.
  • Applicable for calibration, quantitative assays, and method validation.