Learn More
Medchemexpress LLC HY-76847 100mg Medchemexpress, Chenodeoxycholic Acid CDCA CAS:474-25-9 Purity:98%
SDP
Catalog No. 501639061
Encompass_Preferred
Medchemexpress,HY-76847 100mg Chenodeoxycholic Acid CDCA CAS:474-25-9 Formula:C24H40O4 Chenodeoxycholic Acid is a hydrophobic primary bile acid that activates nuclear receptors (FXR) involved in cholesterol metabolism. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-9061 Supplier Medchemexpress LLC Supplier No. HY76847100MG
Medchemexpress,HY-76847 100mg Chenodeoxycholic Acid CDCA CAS:474-25-9 Formula:C24H40O4 Chenodeoxycholic Acid is a hydrophobic primary bile acid that activates nuclear receptors (FXR) involved in cholesterol metabolism. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.