Learn More
AdipoGen Chenodeoxycholic acid (500 mg)
SDP
Supplier:  AdipoGen AGCN20410M500
Encompass_Preferred
Chemical. CAS: 474-25-9. Formula: C24H40O4. MW: 392.6. Synthetic. Originally isolated from bile. Cytotoxic hydrophobic primary bile acid. Activator of farnesoid X receptor (FXR), a nuclear receptor that is hepatoprotective and regulates bile acid synthesis (cholesterol 7alpha-hydroxylase (CYP7A1) suppression), conjugation and transport, as well as genes involved in lipid and glucose metabolism and. Bile acid-controlled signaling pathways are promising novel targets to treat such metabolic diseases as obesity, type II diabetes, insulin resistance, hyperlipidemia and atherosclerosis. Inhibitor of 5beta-reductase (AKR1D1). Potent selective inhibitor of DD2 (AKR1C2). Potent inhibitor of 11beta-HSD1 dehydrogenase. Changes tumor cell viability via IL-6 pathway. Anticancer compound. Apoptosis inducer. Immunosuppressive and anti-inflammatory compound. Modulates oxidative stress. Differentiation regulator of mouse embryonic stem cells. Used for dissolution of cholesterol gallstones.
Catalog No. 50-205-3579