Learn More
Selleck Chemical LLC Chenodeoxycholic Acid 50mg 474-25-9 Chenodiol
SDP
Supplier:  Selleck Chemical LLC S184350MG
Encompass_Preferred
Chenodeoxycholic Acid (Chenodiol) is a naturally occurring human bile acid, and inhibits production of cholesterol in the liver and absorption in the intestines. Chenodeoxycholic Acid is a hydrophobic primary bile acid that activates nuclear receptors (FXR) involved in cholesterol metabolism. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-601-0