Learn More
Cayman Chemical ChenodeoxycholIc AcId-d4 10mg
SDP
Catalog No. 502981619
Encompass_Preferred
An internal standard for the quantification of chenodeoxycholic acid by GC- or LC-MS
Catalog No. 50-298-1619 Supplier Cayman Chemical Supplier No. 2084810 MG
An internal standard for the quantification of chenodeoxycholic acid by GC- or LC-MS