Learn More
Cayman Chemical ChenodeoxycholIc AcId-d4 1mg
SDP
Catalog No. 502981581
Encompass_Preferred
An internal standard for the quantification of chenodeoxycholic acid by GC- or LC-MS
Catalog No. 50-298-1581 Supplier Cayman Chemical Supplier No. 208481 MG
An internal standard for the quantification of chenodeoxycholic acid by GC- or LC-MS