Learn More
Medchemexpress LLC Chenodeoxycholic Acid-d4 | 99102-69-9 | 99.9% | C24H36D4O4 | 1 MG
SDP
Catalog No. 5000433770
Encompass_Preferred
Chenodeoxycholic Acid-d4 is the deuterium labeled Chenodeoxycholic Acid. It is a hydrophobic primary bile acid that activates nuclear receptors (FXR) involved in cholesterol metabolism.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-33770 Supplier Medchemexpress LLC Supplier No. HY76847S1MG
Add to Cart
Add to Cart
Chenodeoxycholic Acid-d4 is the deuterium labeled Chenodeoxycholic Acid. It is a hydrophobic primary bile acid that activates nuclear receptors (FXR) involved in cholesterol metabolism.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.