Learn More
Medchemexpress LLC Chenodeoxycholic acid-d5 | 52840-12-7 | 99.8% | C24H35D5O4 | 1 MG
SDP
Catalog No. 5000434494
Encompass_Preferred
Chenodeoxycholic acid-d5 is the deuterium-labeled version of Chenodeoxycholic Acid, a hydrophobic primary bile acid that activates nuclear receptors (FXR) involved in cholesterol metabolism. This isotope-labeled compound serves as a tracer for quantitation during drug development, leveraging deuteration's potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium-labeled compound
  • Used as a tracer for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
  • Targets FXR pathway
  • Targets Autophagy pathway
  • Targets Endogenous Metabolite pathways
Catalog No. 50-004-34494 Supplier Medchemexpress LLC Supplier No. HY76847S31MG
Add to Cart
Add to Cart
Chenodeoxycholic acid-d5 is the deuterium-labeled version of Chenodeoxycholic Acid, a hydrophobic primary bile acid that activates nuclear receptors (FXR) involved in cholesterol metabolism. This isotope-labeled compound serves as a tracer for quantitation during drug development, leveraging deuteration's potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium-labeled compound
  • Used as a tracer for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
  • Targets FXR pathway
  • Targets Autophagy pathway
  • Targets Endogenous Metabolite pathways