Learn More
Medchemexpress LLC Chenodeoxycholic acid-d5 | 52840-12-7 | 99.8% | C24H35D5O4 | 5 MG
SDP
Catalog No. 5000434087
Encompass_Preferred
Chenodeoxycholic acid-d5 is the deuterium labeled Chenodeoxycholic Acid. Chenodeoxycholic Acid is a hydrophobic primary bile acid that activates nuclear receptors (FXR) involved in cholesterol metabolism.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Activates nuclear receptors (FXR) involved in cholesterol metabolism
  • Deuterium labeling may affect pharmacokinetic and metabolic profiles
Catalog No. 50-004-34087 Supplier Medchemexpress LLC Supplier No. HY76847S35MG
Add to Cart
Add to Cart
Chenodeoxycholic acid-d5 is the deuterium labeled Chenodeoxycholic Acid. Chenodeoxycholic Acid is a hydrophobic primary bile acid that activates nuclear receptors (FXR) involved in cholesterol metabolism.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Activates nuclear receptors (FXR) involved in cholesterol metabolism
  • Deuterium labeling may affect pharmacokinetic and metabolic profiles