Learn More
Medchemexpress LLC Chenodeoxycholic acid sodium | 2646-38-0 | 98.6% | C24H39NaO4 | 500 MG
SDP
Catalog No. 5000434015
Encompass_Preferred
Chenodeoxycholic acid sodium is a hydrophobic primary bile acid that activates nuclear receptors (FXR) involved in cholesterol metabolism. It is for research use only.
  • Activates nuclear receptors (FXR)
  • Involved in cholesterol metabolism
  • Hydrophobic primary bile acid
  • Solid, white to off-white appearance
  • Classified as a steroid
  • Endogenous metabolite
Catalog No. 50-004-34015 Supplier Medchemexpress LLC Supplier No. HY76847A500MG
Add to Cart
Add to Cart
Chenodeoxycholic acid sodium is a hydrophobic primary bile acid that activates nuclear receptors (FXR) involved in cholesterol metabolism. It is for research use only.
  • Activates nuclear receptors (FXR)
  • Involved in cholesterol metabolism
  • Hydrophobic primary bile acid
  • Solid, white to off-white appearance
  • Classified as a steroid
  • Endogenous metabolite